N-(2-Chloroethyl)-N'-[4-(methylsulphanyl)phenyl]urea

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: N-(2-Chloroethyl)-N'-[4-(methylthio)phenyl]urea

OR12062-1G
APOSOR12062-1GEA 132 EUR
APOSOR12062-1G
N-(2-Chloroethyl)-N'-[4-(methylsulphanyl)phenyl]urea
N-(2-Chloroethyl)-N'-[4-(methylsulphanyl)phenyl]urea
Formule: C₁₀H₁₃ClN₂OS
MW: 244.74 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08689728
CAS nummer: 13908-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR