5-Methylthiophene-2-carbonyl chloride

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR12300-1G OR12300-250MG OR12300-100MG OR12300-500MG OR12300-2.5G OR12300-5G
APOSOR12300-1GEA 263 EUR
APOSOR12300-1G APOSOR12300-250MG APOSOR12300-100MG APOSOR12300-500MG APOSOR12300-2.5G APOSOR12300-5G
5-Methylthiophene-2-carbonyl chloride
5-Methylthiophene-2-carbonyl chloride
Formule: C₆H₅ClOS
Kookpunt: 120 °C/8mmHg
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00130092
CAS nummer: 31555-59-6
UN: 3265
ADR: 8,II

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR