5-Bromoindole 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR1278-100G OR1278-500G
APOSOR1278-100GEA 32.2 EUR
APOSOR1278-100G APOSOR1278-500G
5-Bromoindole 99%
5-Bromoindole
Formule: C₈H₆BrN
Smeltpunt: 90…92 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00005670
CAS nummer: 10075-50-0
EINECS: 233-208-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR