3,5-Dimethylisoxazole-4-carbonyl chloride 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR1284-5G OR1284-1G
APOSOR1284-5GEA 146 EUR
APOSOR1284-5G APOSOR1284-1G
3,5-Dimethylisoxazole-4-carbonyl chloride 95%
3,5-Dimethylisoxazole-4-carbonyl chloride
Formule: C₆H₆ClNO₂
Kookpunt: 90 °C/15mmHg
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00052555
CAS nummer: 31301-45-8
UN: 1760
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR