Bis(4-bromophenyl)acetylene

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR13096-1G OR13096-25G OR13096-5G
APOSOR13096-1GEA 22 EUR
APOSOR13096-1G APOSOR13096-25G APOSOR13096-5G
Bis(4-bromophenyl)acetylene
Bis(4-bromophenyl)acetylene
Formule: C₁₄H₈Br₂
Smeltpunt: 183…187 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00185147
CAS nummer: 2789-89-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR