(±)-α-Methoxyphenylacetic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR13894-1G OR13894-500MG OR13894-5G
APOSOR13894-1GEA 24.9 EUR
APOSOR13894-1G APOSOR13894-500MG APOSOR13894-5G
(±)-α-Methoxyphenylacetic acid
(±)-α-Methoxyphenylacetic acid
Formule: C₉H₁₀O₃
Kookpunt: 165 °C/18mmHg
Smeltpunt: 69…71 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064217
CAS nummer: 7021-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR