3-(Morpholinomethyl)phenylboronic acid pinacol ester 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR1396-1G OR1396-250MG
APOSOR1396-1GEA 32.2 EUR
APOSOR1396-1G APOSOR1396-250MG
3-(Morpholinomethyl)phenylboronic acid pinacol ester 97%
3-(Morpholinomethyl)phenylboronic acid pinacol ester
Formule: C₁₇H₂₆BNO₃
Smeltpunt: 50…53 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06797489
CAS nummer: 364794-80-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR