4'-Hydroxybiphenyl-4-carboxylic acid 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR13999-25G OR13999-5G OR13999-100G
APOSOR13999-25GEA 61.4 EUR
APOSOR13999-25G APOSOR13999-5G APOSOR13999-100G
4'-Hydroxybiphenyl-4-carboxylic acid 95%
4'-Hydroxybiphenyl-4-carboxylic acid
Formule: C₁₃H₁₀O₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00059078
CAS nummer: 58574-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR