3-Bromoanthranilic acid 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR14301-5G OR14301-100G OR14301-25G
APOSOR14301-5GEA 22 EUR
APOSOR14301-5G APOSOR14301-100G APOSOR14301-25G
3-Bromoanthranilic acid 97%
3-Bromoanthranilic acid
Formule: C₇H₆BrNO₂
Kookpunt: 343…344 °C
Smeltpunt: 174…178 °C
Dichtheid: 1,793 g/cm³ (25 °C)
Vlampunt: 161 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03618453
CAS nummer: 20776-51-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR