Quinhydrone

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Benzoquinone-hydroquinone, p-Benzoquinone hydroquinone complex, 2,5-Cyclohexadiene-1,4-dione, compd. with 1,4-benzenediol (1:1)

OR14340-100G OR14340-250G OR14340-500G
APOSOR14340-100GEA 52.6 EUR
APOSOR14340-100G APOSOR14340-250G APOSOR14340-500G
Quinhydrone
Quinhydrone
Formule: C₆H₄(OH)₂·C₆H₄O₂
MW: 218.21 g/mol
Smeltpunt: 173…174 °C
Dichtheid: 1.4 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010310
CAS nummer: 106-34-3
EINECS: 203-387-6
UN: 2811
ADR: 6.1 III
Merck Index: 13,08148

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR