5-Bromo-1H-indazole-3-carboxylic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR14502-1G OR14502-250MG OR14502-5G
APOSOR14502-1GEA 67.2 EUR
APOSOR14502-1G APOSOR14502-250MG APOSOR14502-5G
5-Bromo-1H-indazole-3-carboxylic acid
5-Bromo-1H-indazole-3-carboxylic acid
Formule: C₈H₅BrN₂O₂
Smeltpunt: 290…292 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD05663979
CAS nummer: 1077-94-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR