2-Bromo-6-formylpyridine 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR1460-25G OR1460-100G
APOSOR1460-25GEA 55.5 EUR
APOSOR1460-25G APOSOR1460-100G
2-Bromo-6-formylpyridine 95%
2-Bromo-6-formylpyridine
Formule: C₆H₄BrNO
Smeltpunt: 79…85 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD02683546
CAS nummer: 34160-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR