H-4-Pal-OH·2HCl 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR14725-1G OR14725-250MG OR14725-5G
APOSOR14725-1GEA 132 EUR
APOSOR14725-1G APOSOR14725-250MG APOSOR14725-5G
H-4-Pal-OH·2HCl 98%
H-4-Pal-OH·2HCl
Formule: C₈H₁₀N₂O₂·₂HCl
Smeltpunt: 257…259 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01860883
CAS nummer: 37535-49-2

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR