L-Allylglycine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR14736-5G
APOSOR14736-5GEA 38 EUR
APOSOR14736-5G
L-Allylglycine
L-Allylglycine
Formule: C₅H₉NO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002627
CAS nummer: 16338-48-0

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR