4-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR15225-10G OR15225-1G OR15225-5G
APOSOR15225-10GEA 1700 EUR
APOSOR15225-10G APOSOR15225-1G APOSOR15225-5G
4-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylic acid
4-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylic acid
Formule: C₁₀H₉NO₃S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01569174
CAS nummer: 272437-91-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR