Methyl-4-bromo-1-methyl-1H-pyrrole-2-carboxylate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR15346-5G
APOSOR15346-5GEA 505 EUR
APOSOR15346-5G
Methyl-4-bromo-1-methyl-1H-pyrrole-2-carboxylate
Methyl-4-bromo-1-methyl-1H-pyrrole-2-carboxylate
Formule: C₇H₈BrNO₂
Smeltpunt: 63…65 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08275077
CAS nummer: 1196-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR