3-Bromo-5-(chloromethyl)isoxazole 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR15587-1G OR15587-500MG
APOSOR15587-1GEA 343 EUR
APOSOR15587-1G APOSOR15587-500MG
3-Bromo-5-(chloromethyl)isoxazole 95%
3-Bromo-5-(chloromethyl)isoxazole
Formule: C₄H₃BrClNO
Kookpunt: 110 °C/20mmHg
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD06660137
CAS nummer: 124498-15-3

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR