(R)-(-)-1-Boc-3-hydroxypyrrolidine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR15679-100G OR15679-25G OR15679-5G
APOSOR15679-100GEA 86.2 EUR
APOSOR15679-100G APOSOR15679-25G APOSOR15679-5G
(R)-(-)-1-Boc-3-hydroxypyrrolidine
(R)-(-)-1-Boc-3-hydroxypyrrolidine

Chiral intermediate for pharmaceuticals and agrochemicals.

Formule: C₉H₁₇NO₃
Smeltpunt: 62…65 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01317838
CAS nummer: 109431-87-0
UN: 3077
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR