Methyl 2-(bromomethyl)-5-cyanobenzoate 95+%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR15700-250MG
APOSOR15700-250MGEA 114 EUR
APOSOR15700-250MG
Methyl 2-(bromomethyl)-5-cyanobenzoate 95+%
Methyl 2-(bromomethyl)-5-cyanobenzoate
Formule: C₁₀H₈BrNO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD18251143
CAS nummer: 421551-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR