3-Hydroxy-2-nitro-6-picoline 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR16078-100G OR16078-5G OR16078-25G
APOSOR16078-100GEA 649 EUR
APOSOR16078-100G APOSOR16078-5G APOSOR16078-25G
3-Hydroxy-2-nitro-6-picoline 95%
3-Hydroxy-2-nitro-6-picoline
Formule: C₆H₆N₂O₃
Smeltpunt: 105…108 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006259
CAS nummer: 15128-90-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR