NVP-AUY922

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 5-[2,4-Dihydroxy-5-isopropylphenyl]-N-ethyl-4-{4-[(morpholin-4-yl)methyl]phenyl}isoxazole-3-carboxam, Luminespib, 5-(2,4-Dihydroxy-5-isopropylphenyl)-N-ethyl-4-(4-(morpholinomethyl)phenyl)isoxazole-3-carboxamide

OR16660-100MG OR16660-25MG
APOSOR16660-100MGEA 798 EUR
APOSOR16660-100MG APOSOR16660-25MG
NVP-AUY922
NVP-AUY922
Formule: C₂₆H₃₁N₃O₅
MW: 465.54 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD14635361
CAS nummer: 747412-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR