2-Phenoxyisonicotinic acid 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR16904-1G OR16904-500MG OR16904-250MG OR16904-100MG OR16904-2.5G OR16904-5G
APOSOR16904-1GEA 250 EUR
APOSOR16904-1G APOSOR16904-500MG APOSOR16904-250MG APOSOR16904-100MG APOSOR16904-2.5G APOSOR16904-5G
2-Phenoxyisonicotinic acid 97%
2-Phenoxyisonicotinic acid
Formule: C₁₂H₉NO₃
Smeltpunt: 240…242 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09414712
CAS nummer: 51362-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR