2-Bromo-6-nitroanisole

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR17600-5G
APOSOR17600-5GEA 22 EUR
APOSOR17600-5G
2-Bromo-6-nitroanisole
2-Bromo-6-nitroanisole
Formule: C₇H₆BrNO₃
Smeltpunt: 64…65 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09261208
CAS nummer: 98775-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR