3-Bromo-2-methoxyaniline

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR17601-1G OR17601-5G OR17601-10G
APOSOR17601-1GEA 22 EUR
APOSOR17601-1G APOSOR17601-5G APOSOR17601-10G
3-Bromo-2-methoxyaniline
3-Bromo-2-methoxyaniline
Formule: C₇H₈BrNO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD13186737
CAS nummer: 116557-46-1
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR