1,3,5-Tri-tert-butylbenzene 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR17927-100G
APOSOR17927-100GEA 117 EUR
APOSOR17927-100G
1,3,5-Tri-tert-butylbenzene 98%
1,3,5-Tri-tert-butylbenzene
Formule: C₁₈H₃₀
MW: 246.44 g/mol
Kookpunt: 121…122 °C/12mmHg
Smeltpunt: 67…73 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008831
CAS nummer: 1460-02-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR