Phthalaldehyde 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 1,2-Phthalic dicarboxaldehyde, Phthaldialdehyde, o-Phthalaldehyde, OPA, Phthaldicarboxaldehyde, Benzene-1,2-dicarboxaldehyde

OR18349-250G OR18349-25G OR18349-100G
APOSOR18349-250GEA 184 EUR
APOSOR18349-250G APOSOR18349-25G APOSOR18349-100G
Phthalaldehyde 98%
Phthalaldehyde
Formule: C₈H₆O₂
Kookpunt: 83 °C/0,8mmHg
Smeltpunt: 53…57 °C
Dichtheid: 1,13 g/cm³ (25 °C)
Vlampunt: 132 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00003335
CAS nummer: 643-79-8
EINECS: 211-402-2
UN: 2923
ADR: 8+6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR