Toluene-4-sulphonamide 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: PTSA, p-Toluenesulphonamide, 4-Methylbenzenesulphonamide, 4-Toluenesulphonamide

OR18741-500G OR18741-100G OR18741-1KG
APOSOR18741-500GEA 65.8 EUR
APOSOR18741-500G APOSOR18741-100G APOSOR18741-1KG
Toluene-4-sulphonamide 98%
Toluene-4-sulphonamide
Formule: C₇H₉NO₂S
MW: 171.22 g/mol
Kookpunt: 221 °C/10mmHg
Smeltpunt: 136…140 °C
Vlampunt: 202 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011692
CAS nummer: 70-55-3
EINECS: 200-741-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR