9,10-Dichloroanthracene 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR18858-100G OR18858-25G OR18858-5G
APOSOR18858-100GEA 1340 EUR
APOSOR18858-100G APOSOR18858-25G APOSOR18858-5G
9,10-Dichloroanthracene 98%
9,10-Dichloroanthracene
Formule: C₁₄H₈Cl₂
MW: 247.12 g/mol
Kookpunt: 366
Smeltpunt: 210…215 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001246
CAS nummer: 605-48-1
EINECS: 210-087-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR