2,7-Dibromonaphthalene 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR19537-10G OR19537-5G OR19537-25G
APOSOR19537-10GEA 61.4 EUR
APOSOR19537-10G APOSOR19537-5G APOSOR19537-25G
2,7-Dibromonaphthalene 98%
2,7-Dibromonaphthalene
Formule: C₁₀H₆Br₂
Smeltpunt: 139…142 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00238580
CAS nummer: 58556-75-5
UN: 3077
ADR: 9+EHS,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR