4-(Aminomethyl)benzonitrile hydrochloride

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR20018-100G OR20018-25G OR20018-500G
APOSOR20018-100GEA 58.4 EUR
APOSOR20018-100G APOSOR20018-25G APOSOR20018-500G
4-(Aminomethyl)benzonitrile hydrochloride
4-(Aminomethyl)benzonitrile hydrochloride
Formule: C₈H₉ClN₂
Smeltpunt: 274…279 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01861472
CAS nummer: 15996-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR