trans-3-Chloroacrylic acid 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR2048-1G OR2048-5G
APOSOR2048-1GEA 0 EUR
APOSOR2048-1G APOSOR2048-5G
trans-3-Chloroacrylic acid 99%
trans-3-Chloroacrylic acid
Formule: C₃H₃ClO₂
Smeltpunt: 81…83 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064237
CAS nummer: 2345-61-1
EINECS: 219-070-0
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR