2-Cyano-N-methyl-acetamide

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR21454-10G OR21454-1G OR21454-5G
APOSOR21454-10GEA 39.5 EUR
APOSOR21454-10G APOSOR21454-1G APOSOR21454-5G
2-Cyano-N-methyl-acetamide
2-Cyano-N-methyl-acetamide
Formule: C₄H₆N₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00090119
CAS nummer: 6330-25-2

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR