Methyl 5-chloro-4-(2-methoxy-2-oxoethyl)-2-methyl-1H-pyrrole-3-carboxylate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR21814-1G OR21814-250MG
APOSOR21814-1GEA 385 EUR
APOSOR21814-1G APOSOR21814-250MG
Methyl 5-chloro-4-(2-methoxy-2-oxoethyl)-2-methyl-1H-pyrrole-3-carboxylate
Methyl 5-chloro-4-(2-methoxy-2-oxoethyl)-2-methyl-1H-pyrrole-3-carboxylate
Formule: C₁₀H₁₂ClNO₄
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00179171
CAS nummer: 77978-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR