1-Bromo-4-hexylbenzene 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR21925-100G OR21925-25G
APOSOR21925-100GEA 315 EUR
APOSOR21925-100G APOSOR21925-25G
1-Bromo-4-hexylbenzene 97%
1-Bromo-4-hexylbenzene
Formule: C₁₂H₁₇Br
Kookpunt: 145 °C/0,75mmHg
Dichtheid: 1,18 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00061114
CAS nummer: 23703-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR