2-Chloro-3,4-dimethoxybenzyl alcohol

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR21959-10G OR21959-1G OR21959-5G
APOSOR21959-10GEA 262 EUR
APOSOR21959-10G APOSOR21959-1G APOSOR21959-5G
2-Chloro-3,4-dimethoxybenzyl alcohol
2-Chloro-3,4-dimethoxybenzyl alcohol
Formule: C₉H₁₁ClO₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00210141
CAS nummer: 93983-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR