2-Allyl-3-hydroxybenzaldehyde

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR21984-100MG OR21984-1G
APOSOR21984-100MGEA 29.2 EUR
APOSOR21984-100MG APOSOR21984-1G
2-Allyl-3-hydroxybenzaldehyde
2-Allyl-3-hydroxybenzaldehyde
Formule: C₁₀H₁₀O₂
Smeltpunt: 97…98 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00100646
CAS nummer: 79950-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR