4,6-Di(tert-butyl)benzene-1,3-diol 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 4,6-Di-tert-butylresorcinol, 4,6-Di-tert-butylbenzene-1,3-diol

OR22090-10G OR22090-1G OR22090-5G
APOSOR22090-10GEA 0 EUR
APOSOR22090-10G APOSOR22090-1G APOSOR22090-5G
4,6-Di(tert-butyl)benzene-1,3-diol 95%
4,6-Di(tert-butyl)benzene-1,3-diol
Formule: C₁₄H₂₂O₂
MW: 222.33 g/mol
Smeltpunt: 120…125 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00044513
CAS nummer: 5374-06-1
EINECS: 226-366-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR