Diphenylcyclopropenone

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR22091-1G OR22091-5G OR22091-10G
APOSOR22091-1GEA 22 EUR
APOSOR22091-1G APOSOR22091-5G APOSOR22091-10G
Diphenylcyclopropenone
Diphenylcyclopropenone
Formule: C₁₅H₁₀O
Smeltpunt: 118…122 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001311
CAS nummer: 886-38-4
EINECS: 212-948-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR