(4-Ethoxyphenyl)urea

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR22579-1G OR22579-5G OR22579-25G
APOSOR22579-1GEA 58.4 EUR
APOSOR22579-1G APOSOR22579-5G APOSOR22579-25G
(4-Ethoxyphenyl)urea
(4-Ethoxyphenyl)urea
Formule: C₉H₁₂N₂O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00025431
CAS nummer: 150-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR