4-(1H-Pyrazol-1-yl)benzoyl chloride

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR23289-1G OR23289-250MG
APOSOR23289-1GEA 222 EUR
APOSOR23289-1G APOSOR23289-250MG
4-(1H-Pyrazol-1-yl)benzoyl chloride
4-(1H-Pyrazol-1-yl)benzoyl chloride
Formule: C₁₀H₇ClN₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD02681936
CAS nummer: 220461-83-6
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR