4-(4-Methylpiperazin-1-yl)benzoic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR23308-1G OR23308-250MG OR23308-5G
APOSOR23308-1GEA 24.9 EUR
APOSOR23308-1G APOSOR23308-250MG APOSOR23308-5G
4-(4-Methylpiperazin-1-yl)benzoic acid
4-(4-Methylpiperazin-1-yl)benzoic acid
Formule: C₁₂H₁₆N₂O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD02682063
CAS nummer: 86620-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR