2-(4-Morpholino)benzoic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR23383-1G OR23383-25G OR23383-5G
APOSOR23383-1GEA 87.6 EUR
APOSOR23383-1G APOSOR23383-25G APOSOR23383-5G
2-(4-Morpholino)benzoic acid
2-(4-Morpholino)benzoic acid
Formule: C₁₁H₁₃NO₃
Smeltpunt: 151…159 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00527976
CAS nummer: 42106-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR