5-Amino-4,6-dichloro-2-methylpyrimidine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR24758-25G OR24758-5G
APOSOR24758-25GEA 33.7 EUR
APOSOR24758-25G APOSOR24758-5G
5-Amino-4,6-dichloro-2-methylpyrimidine
5-Amino-4,6-dichloro-2-methylpyrimidine
Formule: C₅H₅Cl₂N₃
Smeltpunt: 70…75 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00194053
CAS nummer: 39906-04-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR