2-Amino-4-(4-pyridyl)thiazole

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR2532-1G OR2532-5G
APOSOR2532-1GEA 46.8 EUR
APOSOR2532-1G APOSOR2532-5G
2-Amino-4-(4-pyridyl)thiazole
2-Amino-4-(4-pyridyl)thiazole
Formule: C₈H₇N₃S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00490009
CAS nummer: 30235-28-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR