Potassium methyl malonate 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR25586-100G OR25586-250G OR25586-500G
APOSOR25586-100GEA 35.1 EUR
APOSOR25586-100G APOSOR25586-250G APOSOR25586-500G
Potassium methyl malonate 98%
Potassium methyl malonate
Formule: C₄H₅KO₄
Smeltpunt: 198…205 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00014021
CAS nummer: 38330-80-2
EINECS: 253-886-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR