3-Cyclopentyloxy-4-methoxybenzoic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR25693-10G OR25693-1G
APOSOR25693-10GEA 0 EUR
APOSOR25693-10G APOSOR25693-1G
3-Cyclopentyloxy-4-methoxybenzoic acid
3-Cyclopentyloxy-4-methoxybenzoic acid
Formule: C₁₃H₁₆O₄
Smeltpunt: 160…164 °C
Bewaar temperatuur: Kamertemperatuur
CAS nummer: 144036-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR