Isopropoxybenzene

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR25710-100G OR25710-25G OR25710-5G
APOSOR25710-100GEA 190 EUR
APOSOR25710-100G APOSOR25710-25G APOSOR25710-5G
Isopropoxybenzene
Isopropoxybenzene
Formule: C₉H₁₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00048288
CAS nummer: 2741-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR