5-Chloro-2-hydroxybenzyl alcohol

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 4-Chloro-2-(hydroxymethyl)phenol

OR25788-10G OR25788-1G
APOSOR25788-10GEA 238 EUR
APOSOR25788-10G APOSOR25788-1G
5-Chloro-2-hydroxybenzyl alcohol
5-Chloro-2-hydroxybenzyl alcohol
Formule: C₇H₇ClO₂
MW: 158.59 g/mol
Smeltpunt: 84…87 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00021967
CAS nummer: 5330-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR