1,3-Adamantanediol

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR26342-100G OR26342-25G OR26342-500G
APOSOR26342-100GEA 87.6 EUR
APOSOR26342-100G APOSOR26342-25G APOSOR26342-500G
1,3-Adamantanediol
1,3-Adamantanediol
Formule: C₁₀H₁₆O₂
Smeltpunt: 260 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00154013
CAS nummer: 5001-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR