Diethyl 2-heptylmalonate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR27453-1G OR27453-100MG OR27453-250MG OR27453-500MG OR27453-2.5G OR27453-5G
APOSOR27453-1GEA 409 EUR
APOSOR27453-1G APOSOR27453-100MG APOSOR27453-250MG APOSOR27453-500MG APOSOR27453-2.5G APOSOR27453-5G
Diethyl 2-heptylmalonate
Diethyl 2-heptylmalonate
Formule: C₁₄H₂₆O₄
MW: 258.354 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00027827
CAS nummer: 607-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR